Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173491.3 | NP_775762.1 | 4 | 1028 | ||
GenBank mRNA | AK095592.1 | - | 4 | 978 | ||
BC051353.1 | - | 4 | 951 | |||
BC126449.1 | - | 4 | 1028 | 8992165 |