Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271862.1 | NP_001258791.1 | 17, 18 | 1485 | ||
NM_173516.2 | NP_775787.1 | 17, 18 | 1584 | |||
XM_011535491.2 | XP_011533793.1 | 17, 18 | 1511 | |||
XM_011535493.2 | XP_011533795.1 | 12, 13 | 1179 | |||
XM_017010312.1 | XP_016865801.1 | 17, 18 | 2273 | |||
XM_017010313.1 | XP_016865802.1 | 15, 16 | 2159 | |||
XM_017010314.1 | XP_016865803.1 | 16, 17 | 2129 | |||
XM_017010315.1 | XP_016865804.1 | 15, 16 | 1363 | |||
XM_017010316.1 | XP_016865805.1 | 15, 16 | 1428 | |||
GenBank mRNA | AK093139.1 | - | 17, 18 | 1462 | ||
AK097559.1 | - | 17, 18 | 1564 | |||
BC112246.1 | - | 17, 18 | 1468 | 8327714 |