Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199085.1 | NP_001186014.1 | 2 | 625 | ||
NM_001199089.1 | NP_001186018.1 | 2 | 338 | |||
NM_001199091.1 | NP_001186020.1 | 2 | 466 | |||
NM_173533.3 | NP_775804.2 | 2 | 625 | |||
XM_005244934.1 | XP_005244991.1 | 2 | 410 | |||
XM_005244935.4 | XP_005244992.1 | 2 | 315 | |||
XM_017000473.1 | XP_016855962.1 | 2 | 331 | |||
GenBank mRNA | AK126889.1 | - | 2 | 465 | ||
AK127370.1 | - | 2 | 357 | |||
BC130532.1 | - | 2 | 302 | 40146563 | ||
BC144058.1 | - | 2 | 357 | 9052578 | ||
DB073297.1 | - | 2 | 338 |