Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173587.3 | NP_775858.2 | 2 | 545 | ||
XM_005273932.3 | XP_005273989.1 | 2 | 262 | |||
GenBank mRNA | AB593132.1 | - | 2 | 888 | ||
BC023587.2 | - | 2 | 545 | 4762190 |