Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173587.3 | NP_775858.2 | 10, 11 | 1423 | ||
GenBank mRNA | AB593132.1 | - | 10, 11 | 1766 | ||
BC010608.1 | - | 3, 4 | 381 | |||
BC023587.2 | - | 10, 11 | 1423 | 4762190 |