Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166347.1 | NP_001159819.1 | 18 | 2373 | ||
NM_001166348.1 | NP_001159820.1 | 17 | 2303 | |||
NM_001166349.1 | NP_001159821.1 | 17 | 2173 | |||
NM_173626.3 | NP_775897.3 | 18 | 2370 | |||
XM_006721833.1 | XP_006721896.1 | 18 | 2445 | |||
XM_011524652.1 | XP_011522954.1 | 16 | 2206 | |||
XM_011524653.1 | XP_011522955.1 | 16 | 2134 | |||
XM_011524654.1 | XP_011522956.1 | 16 | 2019 | |||
XM_017024505.1 | XP_016879994.1 | 17 | 2359 | |||
XM_017024507.1 | XP_016879996.1 | 18 | 2419 | |||
GenBank mRNA | AJ544073.1 | - | 18 | 2344 | ||
AK075248.1 | - | 18 | 2333 | |||
AL833468.1 | - | 17 | 2267 | |||
BC015160.1 | - | 2 | 372 | |||
BC035900.1 | - | 18 | 2312 | 5241643 | ||
BC047451.1 | - | 12 | 1422 |