Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174891.3 | NP_777551.2 | 5 | 1148 | ||
GenBank mRNA | AF038188.1 | - | 5 | 712 | ||
AK057249.1 | - | 6 | 1224 | |||
BC003539.1 | - | 4 | 460 | |||
BC011537.1 | - | 4 | 978 | |||
BC025175.1 | - | 5 | 659 | |||
BC137495.1 | - | 5 | 1091 | 9021495 | ||
BC144671.1 | - | 5 | 1077 |