Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003335.2 | NP_003326.2 | 2 | 317 | ||
XM_005265430.2 | XP_005265487.1 | 2 | 301 | |||
XM_006713321.2 | XP_006713384.1 | 2 | 301 | |||
XM_011534070.1 | XP_011532372.1 | 2 | 301 | |||
XM_011534071.1 | XP_011532373.1 | 2 | 301 | |||
XM_011534072.1 | XP_011532374.1 | 2 | 301 | |||
GenBank mRNA | BC006378.2 | - | 2 | 242 | 4299266 | |
BM152341.1 | - | 2 | 319 | |||
BT007026.1 | - | 2 | 146 | |||
L13852.1 | - | 2 | 310 |