Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286367.1 | NP_001273296.1 | 10 | 1737 | ||
NM_174913.2 | NP_777573.1 | 10 | 1903 | |||
XM_005267385.1 | XP_005267442.1 | 10 | 1930 | |||
XM_011536526.1 | XP_011534828.1 | 10 | 1722 | |||
GenBank mRNA | AB095941.1 | - | 10 | 1895 | ||
AK123717.1 | - | 5 | 678 | |||
BC025332.1 | - | 10 | 1715 | 4549328 | ||
BX248018.1 | - | 10 | 1865 | |||
BX647311.1 | - | 10 | 1870 |