Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171186.1 | NP_001164657.1 | 2, 3 | 227 | ||
NM_174951.3 | NP_777611.1 | 2, 3 | 202 | |||
GenBank mRNA | AF494343.1 | - | 2, 3 | 202 | ||
BC112022.1 | - | 2, 3 | 168 | 8327490 | ||
BC113449.1 | - | 2, 3 | 156 | 8322501 | ||
BC143805.1 | - | 2, 3 | 227 | 9052320 |