Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161368.1 | NP_001154840.1 | 12 | 2023 | ||
NM_174977.3 | NP_777637.1 | 12 | 2391 | |||
XM_006724235.2 | XP_006724298.1 | 12 | 2407 | |||
XM_011530164.2 | XP_011528466.1 | 12 | 2571 | |||
XM_017028787.1 | XP_016884276.1 | 11 | 2520 | |||
XM_017028788.1 | XP_016884277.1 | 10 | 2437 | |||
XM_017028789.1 | XP_016884278.1 | 11 | 2193 | |||
XM_017028791.1 | XP_016884280.1 | 11 | 2220 | |||
GenBank mRNA | AK092002.1 | - | 13 | 2440 | ||
AK093418.1 | - | 12 | 2452 | |||
AK093448.1 | - | 12 | 2375 |