Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174978.2 | NP_777638.2 | 18 | 1883 | ||
XM_011536690.2 | XP_011534992.1 | 19 | 2013 | |||
XM_017021247.1 | XP_016876736.1 | 19 | 2011 | |||
XM_017021248.1 | XP_016876737.1 | 17 | 1846 | |||
XM_017021249.1 | XP_016876738.1 | 15 | 1578 | |||
XM_017021250.1 | XP_016876739.1 | 14 | 1928 | |||
XM_017021251.1 | XP_016876740.1 | 14 | 1529 | |||
GenBank mRNA | AK098187.1 | - | 18 | 1868 | ||
AL832219.1 | - | 18 | 1933 | |||
BC125070.1 | - | 18 | 1854 | 40117603 | ||
EB388156.1 | - | 3 | 344 |