Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206955.1 | NP_001193884.1 | 23 | 3340 | ||
NM_001206956.1 | NP_001193885.1 | 15 | 2059 | |||
NM_175607.2 | NP_783200.1 | 24 | 3295 | |||
NM_175613.2 | NP_783302.1 | 15 | 2062 | |||
XM_006713004.3 | XP_006713067.1 | 23 | 3305 | |||
XM_011533425.2 | XP_011531727.1 | 23 | 3308 | |||
XM_011533426.2 | XP_011531728.1 | 24 | 3527 | |||
XM_011533427.2 | XP_011531729.1 | 24 | 3508 | |||
XM_011533428.2 | XP_011531730.1 | 22 | 3497 | |||
XM_011533429.2 | XP_011531731.1 | 22 | 3432 | |||
XM_011533430.2 | XP_011531732.1 | 22 | 3393 | |||
XM_011533431.2 | XP_011531733.1 | 21 | 6142 | |||
XM_017005782.1 | XP_016861271.1 | 24 | 3250 | |||
XM_017005783.1 | XP_016861272.1 | 25 | 4124 | |||
XM_017005784.1 | XP_016861273.1 | 23 | 3585 | |||
XM_017005785.1 | XP_016861274.1 | 24 | 3232 | |||
XM_017005786.1 | XP_016861275.1 | 22 | 3494 | |||
XM_017005787.1 | XP_016861276.1 | 22 | 3390 | |||
GenBank mRNA | AF464063.1 | - | 5 | 1206 | ||
AF549455.1 | - | 21 | 3338 | |||
AK057278.1 | - | 15 | 2040 | |||
AK097633.1 | - | 18 | 2586 | |||
AK314396.1 | - | 15 | 2040 | |||
AY090737.1 | - | 23 | 3177 | |||
BC026119.1 | - | 15 | 2042 | 4826091 |