Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300795.1 | NP_001287724.1 | 5 | 325 | ||
NM_003340.6 | NP_003331.1 | 5 | 632 | |||
NM_181886.3 | NP_871615.1 | 5 | 840 | |||
NM_181887.2 | NP_871616.1 | 5 | 610 | |||
NM_181888.3 | NP_871617.1 | 5 | 489 | |||
NM_181889.2 | NP_871618.1 | 5 | 512 | |||
NM_181890.2 | NP_871619.1 | 5 | 454 | |||
NM_181891.2 | NP_871620.1 | 5 | 797 | |||
NM_181892.3 | NP_871621.1 | 4 | 822 | |||
NM_181893.2 | NP_871622.1 | 4 | 302 | |||
XM_017008584.1 | XP_016864073.1 | 4 | 664 | |||
GenBank mRNA | AK095822.1 | - | 4 | 737 | ||
AK127304.1 | - | 3 | 3245 | |||
AK315775.1 | - | 5 | 398 | |||
AL833414.1 | - | 5 | 416 | |||
AU131013.1 | - | 5 | 400 | |||
BC003395.1 | - | 5 | 312 | 3447673 | ||
BC037894.2 | - | 5 | 609 | 5275356 | ||
BC066917.1 | - | 5 | 343 | 4792462 | ||
BM541512.1 | - | 5 | 594 | |||
BQ428985.1 | - | 5 | 307 | |||
BU567649.1 | - | 5 | 482 | |||
BU570661.1 | - | 4 | 257 | |||
CR456916.1 | - | 4 | 147 | |||
DA335600.1 | - | 5 | 390 | |||
DA550074.1 | - | 5 | 244 | |||
DA808523.1 | - | 5 | 565 | |||
DA965437.1 | - | 4 | 308 | |||
DB234695.1 | - | 4 | 246 | |||
HY109217.1 | - | 5 | 454 | |||
U39318.1 | - | 4 | 192 |