Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202489.1 | NP_001189418.1 | 4 | 671 | ||
NM_003343.5 | NP_003334.2 | 6 | 753 | |||
NM_182688.2 | NP_872630.1 | 7 | 868 | |||
XM_017028469.1 | XP_016883958.1 | 6 | 712 | |||
GenBank mRNA | AF032456.1 | - | 6 | 720 | ||
AK075143.1 | - | 2 | 894 | |||
AK122700.1 | - | 2 | 2170 | |||
AK290629.1 | - | 6 | 752 | |||
AL834129.1 | - | 2 | 1487 | |||
BC001738.1 | - | 6 | 716 | 3536853 | ||
BC008351.1 | - | 6 | 713 | 3536171 | ||
BC011569.1 | - | 6 | 725 | 4548763 |