Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079537.1 | NP_001073005.1 | 6 | 838 | ||
NM_177452.3 | NP_803235.1 | 5 | 754 | |||
XM_011536409.2 | XP_011534711.1 | 7 | 858 | |||
GenBank mRNA | AK056690.1 | - | 5 | 724 | ||
AK294339.1 | - | 4 | 536 | |||
AK310730.1 | - | 6 | 648 | |||
AK315900.1 | - | 6 | 766 | |||
AL833179.1 | - | 6 | 722 | |||
BC047328.1 | - | 5 | 638 | 5534151 | ||
BX161510.1 | - | 6 | 730 | |||
BX248013.1 | - | 6 | 794 |