Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178229.4 | NP_839943.2 | 9 | 938 | ||
XM_011509198.2 | XP_011507500.1 | 9 | 927 | |||
XM_017000317.1 | XP_016855806.1 | 9 | 927 | |||
XM_017000318.1 | XP_016855807.1 | 7 | 681 | |||
GenBank mRNA | AB105103.1 | - | 9 | 874 | ||
AY253300.1 | - | 9 | 863 | |||
BC131536.1 | - | 9 | 896 | 40082932 |