Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSS51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024593.1 | NP_001019764.1 | 4 | 514 | ||
GenBank mRNA | AK096884.1 | - | 4 | 515 | ||
AK309870.1 | - | 5 | 570 | |||
BC094693.1 | - | 4 | 493 | 30529259 | ||
BC109255.1 | - | 3 | 427 | |||
BC109256.1 | - | 3 | 427 | |||
BX537678.1 | - | 4 | 565 | |||
CR936824.1 | - | 4 | 496 |