Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003356.3 | NP_003347.1 | 4 | 824 | ||
NM_022803.2 | NP_073714.1 | 4 | 824 | |||
GenBank mRNA | AF001787.1 | - | 4 | 652 | ||
AF011449.1 | - | 3 | 567 | |||
AK092685.1 | - | 4 | 471 | |||
AK290630.1 | - | 4 | 672 | |||
AK291889.1 | - | 4 | 638 | |||
AK309873.1 | - | 4 | 672 | |||
KU178372.1 | - | 3 | 469 | |||
U82818.1 | - | 4 | 622 | |||
U84763.1 | - | 4 | 622 |