Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEZ6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098635.1 | NP_001092105.1 | 8 | 1804 | ||
NM_001290202.1 | NP_001277131.1 | 8 | 1640 | |||
NM_178860.4 | NP_849191.3 | 8 | 1804 | |||
XM_011524315.1 | XP_011522617.1 | 8 | 1804 | |||
XM_011524317.2 | XP_011522619.1 | 8 | 1804 | |||
GenBank mRNA | AF502129.1 | - | 8 | 1892 | ||
AK055383.1 | - | 2 | 76 | |||
AK091522.1 | - | 8 | 1640 | |||
AK125377.1 | - | 8 | 1766 | |||
AK223620.1 | - | 8 | 1766 | |||
AL834405.1 | - | 7 | 1549 | |||
AY038048.1 | - | 8 | 1795 |