Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145017.1 | NP_001138489.1 | 8 | 1279 | ||
NM_181774.3 | NP_861439.3 | 7 | 1156 | |||
XM_011537627.2 | XP_011535929.1 | 7 | 1078 | |||
XM_011537629.2 | XP_011535931.1 | 7 | 1156 | |||
XM_011537630.1 | XP_011535932.1 | 5 | 503 | |||
XM_011537631.1 | XP_011535933.1 | 5 | 514 | |||
XM_011537633.1 | XP_011535935.1 | 3 | 366 | |||
XM_011537634.2 | XP_011535936.1 | 7 | 1112 | |||
GenBank mRNA | AK127978.1 | - | 7 | 1149 | ||
AK131483.1 | - | 6 | 2637 | |||
AY162215.1 | - | 7 | 974 | |||
BC101092.1 | - | 8 | 1002 | 40009824 | ||
BC101093.1 | - | 8 | 1002 | 40009826 | ||
BC101094.1 | - | 7 | 879 | 40009832 | ||
BC101095.2 | - | 7 | 879 | 40009834 |