Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321116.1 | NP_001308045.1 | 3 | 4582 | ||
NM_001321117.1 | NP_001308046.1 | 3 | 4571 | |||
NM_181846.2 | NP_862829.1 | 3 | 4587 | |||
XM_006723192.3 | XP_006723255.1 | 3 | 4559 | |||
XM_011526917.2 | XP_011525219.1 | 3 | 5665 | |||
XM_011526918.2 | XP_011525220.1 | 2 | 4105 | |||
XM_017026746.1 | XP_016882235.1 | 3 | 4585 | |||
XM_017026747.1 | XP_016882236.1 | 3 | 4538 | |||
GenBank mRNA | AK128716.1 | - | 3 | 4579 | ||
BX537741.1 | - | 3 | 4591 | |||
BX647879.1 | - | 1 | 2925 |