Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL9L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182557.2 | NP_872363.1 | 8 | 7172 | ||
XM_005271511.2 | XP_005271568.1 | 10 | 7172 | |||
XM_006718815.2 | XP_006718878.1 | 9 | 6670 | |||
XM_017017572.1 | XP_016873061.1 | 10 | 7421 | |||
GenBank mRNA | AB094091.1 | - | 8 | 7173 |