Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153453.1 | NP_703154.1 | 3 | 1253 | ||
NM_182645.3 | NP_872586.1 | 4 | 1775 | |||
XM_005266883.2 | XP_005266940.1 | 4 | 1737 | |||
GenBank mRNA | AJ578053.1 | - | 4 | 1759 | ||
AY056583.1 | - | 3 | 1253 |