Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017415.1 | NP_001017415.1 | 2 | 429 | ||
NM_001017416.1 | NP_001017416.1 | 2 | 334 | |||
NM_003368.4 | NP_003359.3 | 2 | 859 | |||
GenBank mRNA | AB014458.1 | - | 2 | 210 | ||
AB208893.1 | - | 2 | 491 | |||
AF117386.1 | - | 2 | 346 | |||
AL117575.1 | - | 1 | 181 | |||
AL599240.1 | - | 2 | 341 | |||
BC018745.1 | - | 2 | 859 | |||
BC032364.1 | - | 2 | 509 | |||
BC050525.1 | - | 2 | 563 | 6473568 |