Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113498.2 | NP_001106970.3 | 6 | 1147 | ||
NM_182830.4 | NP_878250.2 | 6 | 853 | |||
NR_103766.1 | 6 | 853 | ||||
XM_011536519.2 | XP_011534821.1 | 6 | 1002 | |||
XM_011536522.2 | XP_011534824.1 | 6 | 1512 | |||
XM_011536525.2 | XP_011534827.1 | 3 | 310 | |||
XM_017021060.1 | XP_016876549.1 | 2 | 113 | |||
XM_017021061.1 | XP_016876550.1 | 6 | 1512 | |||
GenBank mRNA | AK309211.1 | - | 6 | 1150 | ||
AY358125.1 | - | 5 | 727 | |||
AY369208.1 | - | 6 | 943 | |||
BC142947.1 | - | 6 | 896 |