Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113498.2 | NP_001106970.3 | 7 | 1373 | ||
NM_182830.4 | NP_878250.2 | 7 | 1079 | |||
NR_103766.1 | 7 | 1079 | ||||
XM_011536519.2 | XP_011534821.1 | 7 | 1228 | |||
XM_011536522.2 | XP_011534824.1 | 7 | 1738 | |||
XM_011536525.2 | XP_011534827.1 | 4 | 536 | |||
XM_017021060.1 | XP_016876549.1 | 3 | 339 | |||
XM_017021061.1 | XP_016876550.1 | 7 | 1738 | |||
GenBank mRNA | AK309211.1 | - | 7 | 1376 | ||
AY358125.1 | - | 6 | 953 | |||
AY369208.1 | - | 7 | 1169 | |||
BC142947.1 | - | 7 | 1122 |