Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113498.2 | NP_001106970.3 | 15 | 3013 | ||
NM_182830.4 | NP_878250.2 | 15 | 2719 | |||
XM_011536519.2 | XP_011534821.1 | 15 | 2868 | |||
XM_011536525.2 | XP_011534827.1 | 12 | 2150 | |||
XM_017021060.1 | XP_016876549.1 | 11 | 1979 | |||
GenBank mRNA | AK123307.1 | - | 4 | 563 | ||
AK309211.1 | - | 15 | 3016 | |||
AY328482.1 | - | 9 | 1646 | |||
AY358125.1 | - | 14 | 2593 | |||
AY369208.1 | - | 15 | 2809 |