Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301144.1 | NP_001288073.1 | 3 | 664 | ||
NM_080745.4 | NP_542783.2 | 3 | 656 | |||
NM_182985.4 | NP_892030.3 | 4 | 1133 | |||
GenBank mRNA | AF302088.1 | - | 3 | 512 | ||
AK226115.1 | - | 2 | 4920 | |||
AK292252.1 | - | 4 | 977 | |||
AL360161.1 | - | 3 | 499 | |||
AY305385.1 | - | 4 | 831 | |||
BC024199.1 | - | 4 | 951 | |||
BC033314.1 | - | 3 | 452 | 5166812 | ||
BC047945.1 | - | 4 | 970 | 5744224 |