Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164688.1 | NP_001158160.1 | 3 | 3037 | ||
NM_183059.2 | NP_898882.1 | 3 | 3040 | |||
XM_017001151.1 | XP_016856640.1 | 2 | 2407 | |||
GenBank mRNA | AY191519.1 | - | 3 | 3040 | ||
BC065541.1 | - | 3 | 2172 | 6140075 | ||
BX647803.1 | - | 3 | 2256 |