Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194313.2 | NP_919289.2 | 13 | 5889 | ||
XM_005251451.2 | XP_005251508.1 | 9 | 4782 | |||
XM_006716764.2 | XP_006716827.1 | 7 | 4493 | |||
XM_011517861.1 | XP_011516163.1 | 14 | 6110 | |||
XM_011517863.2 | XP_011516165.1 | 13 | 7199 | |||
XM_017014697.1 | XP_016870186.1 | 13 | 6203 | |||
XM_017014698.1 | XP_016870187.1 | 12 | 5976 | |||
GenBank mRNA | AK125180.1 | - | 3 | 2603 | ||
AK125872.1 | - | 4 | 4265 | |||
BC048311.1 | - | 4 | 2636 |