Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052900.2 | NP_443132.3 | 2, 3 | 568 | ||
NM_198123.1 | NP_937756.1 | 2, 3 | 568 | |||
NM_198124.1 | NP_937757.1 | 3, 4 | 460 | |||
XM_011516816.2 | XP_011515118.1 | 2, 3 | 1054 | |||
XM_017013008.1 | XP_016868497.1 | 2, 3 | 1054 | |||
XM_017013009.1 | XP_016868498.1 | 2, 3 | 1054 | |||
XM_017013010.1 | XP_016868499.1 | 2, 3 | 1054 | |||
XM_017013011.1 | XP_016868500.1 | 2, 3 | 1054 | |||
GenBank mRNA | AB114604.1 | - | 2, 3 | 568 | ||
AB114605.1 | - | 3, 4 | 460 | |||
AY210419.1 | - | 2, 3 | 439 | |||
BC140003.1 | - | 2, 3 | 494 |