Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052900.2 | NP_443132.3 | 41 | 6486 | ||
NM_198123.1 | NP_937756.1 | 42 | 6798 | |||
NM_198124.1 | NP_937757.1 | 43 | 6690 | |||
XM_011516815.2 | XP_011515117.1 | 16 | 2299 | |||
XM_011516816.2 | XP_011515118.1 | 41 | 6972 | |||
XM_017013008.1 | XP_016868497.1 | 41 | 7206 | |||
XM_017013009.1 | XP_016868498.1 | 40 | 7074 | |||
XM_017013010.1 | XP_016868499.1 | 40 | 6894 | |||
XM_017013011.1 | XP_016868500.1 | 38 | 6684 | |||
XM_017013012.1 | XP_016868501.1 | 40 | 6257 | |||
GenBank mRNA | AB067481.2 | - | 27 | 4449 | ||
AB114604.1 | - | 42 | 6798 | |||
AB114605.1 | - | 43 | 6690 | |||
AY210419.1 | - | 41 | 6357 |