Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEGFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243733.1 | NP_001230662.1 | 2, 3 | 356 | ||
NM_003377.4 | NP_003368.1 | 2, 3 | 356 | |||
GenBank mRNA | BC008818.2 | - | 2, 3 | 146 | 4053976 | |
U43368.1 | - | 2, 3 | 108 | |||
U43369.1 | - | 2, 3 | 106 | |||
U48801.1 | - | 2, 3 | 109 | |||
U52819.1 | - | 2, 3 | 110 |