Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM120AOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322224.1 | NP_001309153.1 | 3 | 1807 | ||
NM_198841.2 | NP_942138.2 | 3 | 2736 | |||
NR_136229.1 | 3 | 2143 | ||||
NR_136230.1 | 4 | 2264 | ||||
NR_136231.1 | 4 | 2857 | ||||
NR_136232.1 | 4 | 2071 | ||||
NR_136233.1 | 3 | 1894 | ||||
NR_136234.1 | 4 | 1928 | ||||
NR_136235.1 | 3 | 1950 | ||||
NR_136236.1 | 3 | 2153 | ||||
NR_136237.1 | 4 | 2274 | ||||
NR_136238.1 | 4 | 2015 | ||||
GenBank mRNA | AK056096.1 | - | 3 | 2736 | ||
BC063650.1 | - | 3 | 1701 | |||
BC080634.1 | - | 4 | 1798 |