Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270391.1 | NP_001257320.1 | 6 | 739 | ||
NM_001270392.1 | NP_001257321.1 | 5 | 517 | |||
NM_177532.4 | NP_803876.1 | 6 | 739 | |||
NM_201431.2 | NP_958834.1 | 6 | 649 | |||
XM_011531687.2 | XP_011529989.1 | 6 | 641 | |||
XM_011531688.1 | XP_011529990.1 | 7 | 715 | |||
XM_011531689.2 | XP_011529991.1 | 6 | 641 | |||
XM_011531690.2 | XP_011529992.1 | 5 | 3275 | |||
XM_011531691.1 | XP_011529993.1 | 4 | 367 | |||
XM_017007837.1 | XP_016863326.1 | 3 | 2619 | |||
XM_017007838.1 | XP_016863327.1 | 6 | 641 | |||
GenBank mRNA | AK002032.1 | - | 1 | 474 | ||
AK126346.1 | - | 6 | 641 | |||
AY217664.1 | - | 6 | 553 | |||
AY217665.1 | - | 5 | 386 | |||
BC058835.1 | - | 6 | 582 | 6668006 | ||
CR749283.1 | - | 6 | 739 |