Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015458.3 | NP_056273.2 | 1, 2 | 586 | ||
XM_011543830.2 | XP_011542132.1 | 1, 2 | 250 | |||
XM_017013753.1 | XP_016869242.1 | 1, 2 | 544 | |||
GenBank mRNA | AJ297823.1 | - | 1, 2 | 270 | ||
AK000444.1 | - | 1, 2 | 270 | |||
AK223525.1 | - | 1, 2 | 270 | |||
AK303543.1 | - | 1, 2 | 259 | |||
BC022003.1 | - | 1, 2 | 229 | 4825583 | ||
BC034990.2 | - | 1, 2 | 587 | 4831337 | ||
BC093788.1 | - | 1, 2 | 231 | 7939633 | ||
BC101623.1 | - | 1, 2 | 231 | 8069129 | ||
BX538176.1 | - | 1, 2 | 252 |