Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WNK1 SIRNA Assays ›
Gene Symbol:
WNK1
Gene Name:
WNK lysine deficient protein kinase 1
Gene Aliases:
HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65
Chromosome Location:
Chr.12: 752552 - 911452 on Build GRCh38
UniGene ID:
Hs.744906
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
130234
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WNK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184985.1
NP_001171914.1
6
2101
NM_014823.2
NP_055638.2
6
2101
NM_018979.3
NP_061852.3
6
2101
NM_213655.4
NP_998820.3
6
2101
XM_006719003.2
XP_006719066.1
6
2426
XM_011520997.2
XP_011519299.1
6
2472
XM_011520998.2
XP_011519300.1
6
2426
XM_011520999.2
XP_011519301.1
6
2426
XM_011521000.2
XP_011519302.1
6
2426
XM_011521001.2
XP_011519303.1
6
2426
XM_011521002.2
XP_011519304.1
6
2426
XM_011521003.2
XP_011519305.1
6
2426
XM_011521004.2
XP_011519306.1
6
2426
XM_011521005.2
XP_011519307.1
3
910
XM_011521006.2
XP_011519308.1
6
2426
XM_011521007.2
XP_011519309.1
6
2426
XM_011521008.2
XP_011519310.1
6
2426
XM_011521009.2
XP_011519311.1
6
2426
XM_017019834.1
XP_016875323.1
6
2426
XM_017019835.1
XP_016875324.1
6
2426
XM_017019836.1
XP_016875325.1
6
2426
XM_017019837.1
XP_016875326.1
6
2426
XM_017019838.1
XP_016875327.1
6
2426
XM_017019839.1
XP_016875328.1
6
2426
GenBank mRNA
AB002342.2
-
6
1256
AF061944.1
-
6
2101
AJ296290.1
-
6
1458
JQ358908.1
-
6
1458
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of systemic arterial blood pressure
protein phosphorylation
ion transport
negative regulation of phosphatase activity
peptidyl-threonine phosphorylation
positive regulation of ion transmembrane transporter activity
intracellular signal transduction
protein autophosphorylation
neuron development
regulation of cellular process
negative regulation of protein serine/threonine kinase activity
negative regulation of pancreatic juice secretion
regulation of ion homeostasis
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
chloride channel inhibitor activity
potassium channel inhibitor activity
protein kinase binding
phosphatase binding
protein serine/threonine kinase inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0