Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002838.3 | NP_001002838.1 | 7 | 1641 | ||
NM_020922.4 | NP_065973.2 | 7 | 1641 | |||
XM_006724593.3 | XP_006724656.1 | 7 | 1579 | |||
XM_011530802.2 | XP_011529104.1 | 7 | 1579 | |||
XM_017029741.1 | XP_016885230.1 | 7 | 1580 | |||
XM_017029742.1 | XP_016885231.1 | 7 | 1657 | |||
XM_017029743.1 | XP_016885232.1 | 7 | 1572 | |||
XM_017029744.1 | XP_016885233.1 | 7 | 1474 | |||
XM_017029745.1 | XP_016885234.1 | 7 | 1579 | |||
GenBank mRNA | AJ409088.2 | - | 6 | 1202 | ||
AY082340.1 | - | 6 | 1202 | |||
AY352048.1 | - | 7 | 1641 |