Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM26 SIRNA Assays ›
Gene Symbol:
RBM26
Gene Name:
RNA binding motif protein 26
Gene Aliases:
ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17
Chromosome Location:
Chr.13: 79311827 - 79406265 on Build GRCh38
UniGene ID:
Hs.558528
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
130266
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM26
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286631.1
NP_001273560.1
9
1786
NM_001286632.1
NP_001273561.1
9
1771
NM_022118.4
NP_071401.3
9
1771
XM_005266491.4
XP_005266548.1
9
1853
XM_005266497.1
XP_005266554.1
9
1853
XM_006719857.1
XP_006719920.1
9
1853
XM_011535179.1
XP_011533481.1
9
1853
XM_011535180.2
XP_011533482.1
9
1853
XM_011535181.1
XP_011533483.1
9
1853
XM_011535182.1
XP_011533484.1
9
1838
XM_011535183.2
XP_011533485.1
9
1853
XM_011535184.1
XP_011533486.1
9
1823
XM_011535185.2
XP_011533487.1
9
1823
XM_011535186.2
XP_011533488.1
9
1815
XM_011535187.2
XP_011533489.1
9
1823
XM_011535188.2
XP_011533490.1
9
1815
XM_011535189.1
XP_011533491.1
9
1853
XM_011535190.1
XP_011533492.1
9
1853
XM_011535191.2
XP_011533493.1
9
1853
XM_011535192.2
XP_011533494.1
9
1815
XM_011535193.2
XP_011533495.1
9
1808
XM_011535195.1
XP_011533497.1
9
1551
XM_017020688.1
XP_016876177.1
9
1853
XM_017020689.1
XP_016876178.1
9
1838
XM_017020690.1
XP_016876179.1
9
1845
XM_017020691.1
XP_016876180.1
9
1845
XM_017020692.1
XP_016876181.1
9
1815
XM_017020693.1
XP_016876182.1
9
1853
XM_017020694.1
XP_016876183.1
9
1838
XM_017020695.1
XP_016876184.1
9
1838
XM_017020696.1
XP_016876185.1
9
1845
XM_017020697.1
XP_016876186.1
9
1845
XM_017020698.1
XP_016876187.1
9
1808
XM_017020699.1
XP_016876188.1
9
1830
XM_017020700.1
XP_016876189.1
9
1815
XM_017020701.1
XP_016876190.1
9
1853
XM_017020702.1
XP_016876191.1
9
1853
XM_017020703.1
XP_016876192.1
9
1823
GenBank mRNA
AK027456.1
-
3
420
AK302871.1
-
9
1380
AL834343.1
-
4
684
AX775885.1
-
3
420
BC041655.1
-
9
1338
4384399
BC111697.1
-
9
1332
40038080
BC111739.1
-
9
1348
40038082
BX648672.1
-
9
1329
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_134077, AK139077, AK051686, AK049083, BC066051, and others.
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
negative regulation of phosphatase activity
nucleotide binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline