Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320458.1 | NP_001307387.1 | 2, 3 | 395 | ||
NM_003396.2 | NP_003387.1 | 2, 3 | 395 | |||
XM_011525178.2 | XP_011523480.1 | 2, 3 | 505 | |||
GenBank mRNA | AB063483.1 | - | 2, 3 | 379 | ||
AK127615.1 | - | 2, 3 | 395 | |||
AY358217.1 | - | 2, 3 | 415 | |||
BC064534.1 | - | 2, 3 | 369 | 5588904 |