Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171930.1 | NP_001165401.1 | 6 | 782 | ||
NM_001171931.1 | NP_001165402.1 | 6 | 782 | |||
NM_001171932.1 | NP_001165403.1 | 6 | 782 | |||
NM_022124.5 | NP_071407.4 | 6 | 782 | |||
NM_052836.3 | NP_443068.1 | 6 | 782 | |||
XM_006717940.3 | XP_006718003.1 | 6 | 776 | |||
XM_006717942.3 | XP_006718005.1 | 6 | 776 | |||
XM_011540039.2 | XP_011538341.1 | 6 | 776 | |||
XM_011540042.2 | XP_011538344.1 | 6 | 776 | |||
XM_011540043.2 | XP_011538345.1 | 6 | 776 | |||
XM_011540044.2 | XP_011538346.1 | 6 | 417 | |||
XM_011540045.2 | XP_011538347.1 | 6 | 776 | |||
XM_011540048.2 | XP_011538350.1 | 6 | 776 | |||
XM_011540049.2 | XP_011538351.1 | 6 | 776 | |||
XM_011540051.2 | XP_011538353.1 | 6 | 776 | |||
XM_017016499.1 | XP_016871988.1 | 6 | 782 | |||
XM_017016500.1 | XP_016871989.1 | 6 | 776 | |||
GenBank mRNA | AF312024.1 | - | 5 | 392 | ||
AY010111.2 | - | 5 | 392 | |||
AY358617.1 | - | 6 | 432 | |||
BC011570.1 | - | 6 | 787 | |||
BC032581.1 | - | 6 | 788 | |||
BC065284.1 | - | 6 | 759 | |||
BC108254.1 | - | 4 | 622 | |||
BC136976.1 | - | 6 | 652 | |||
BC136977.1 | - | 6 | 651 | |||
BC139903.1 | - | 6 | 774 |