Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022133.3 | NP_071416.2 | 5 | 996 | ||
NM_152836.2 | NP_690049.1 | 4 | 750 | |||
NM_152837.2 | NP_690050.1 | 3 | 663 | |||
XM_005251282.4 | XP_005251339.1 | 5 | 703 | |||
XM_005251283.2 | XP_005251340.1 | 6 | 840 | |||
XM_011517574.2 | XP_011515876.1 | 3 | 93 | |||
GenBank mRNA | AK290903.1 | - | 4 | 666 | ||
AL833763.1 | - | 3 | 570 | |||
BC033630.1 | - | 4 | 658 | 5224216 | ||
CB962009.1 | - | 4 | 728 |