Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166412.1 | NP_001159884.1 | 8 | 915 | ||
NM_022138.2 | NP_071421.1 | 8 | 948 | |||
XM_011536065.1 | XP_011534367.1 | 8 | 946 | |||
XM_011536066.1 | XP_011534368.1 | 8 | 913 | |||
GenBank mRNA | AB014730.1 | - | 8 | 715 | ||
AB014737.1 | - | 8 | 748 | |||
AF173892.1 | - | 7 | 570 | |||
AJ249902.1 | - | 3 | 194 | |||
AJ420521.1 | - | 8 | 895 | |||
AK056700.1 | - | 8 | 842 | |||
AK314613.1 | - | 8 | 882 | |||
AL832303.1 | - | 8 | 850 | |||
BC028420.1 | - | 8 | 918 | 4826774 | ||
BC047583.1 | - | 8 | 887 | 4826950 |