Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003405.3 | NP_003396.1 | 1, 2 | 333 | ||
GenBank mRNA | AK312650.1 | - | 1, 2 | 177 | ||
BC003047.1 | - | 1, 2 | 268 | 3543571 | ||
CR456612.1 | - | 1, 2 | 136 | |||
L20422.1 | - | 1, 2 | 101 | |||
S80794.1 | - | 1, 2 | 283 | |||
X57345.1 | - | 16 | ||||
X78138.1 | - | 1, 2 | 292 |