Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022365.3 | NP_071760.2 | 4 | 709 | ||
XM_011519614.2 | XP_011517916.1 | 4 | 756 | |||
XM_017016536.1 | XP_016872025.1 | 4 | 756 | |||
GenBank mRNA | AK027263.1 | - | 4 | 529 | ||
AK308369.1 | - | 4 | 715 | |||
AY225122.1 | - | 4 | 422 | |||
BC110894.1 | - | 4 | 709 | 5164559 |