Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022371.3 | NP_071766.2 | 6 | 1332 | ||
GenBank mRNA | AJ299403.1 | - | 6 | 1221 | ||
AK025998.1 | - | 6 | 1211 | |||
AK130132.1 | - | 4 | 651 | |||
AK299966.1 | - | 5 | 929 | |||
AK316530.1 | - | 5 | 941 | |||
BC001085.1 | - | 6 | 1230 | 2959819 | ||
BC007571.1 | - | 6 | 1221 | 3029304 | ||
BC011746.1 | - | 6 | 1217 | 3343691 | ||
BC018292.1 | - | 6 | 1222 | 3457259 | ||
BC088368.1 | - | 5 | 959 | |||
CR457354.1 | - | 6 | 1180 |