Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022458.3 | NP_071903.2 | 6 | 656 | ||
XM_005249555.2 | XP_005249612.1 | 6 | 631 | |||
XM_005249558.2 | XP_005249615.1 | 6 | 631 | |||
XM_017012507.1 | XP_016867996.1 | 6 | 631 | |||
XM_017012508.1 | XP_016867997.1 | 6 | 589 | |||
XM_017012509.1 | XP_016867998.1 | 6 | 631 | |||
XM_017012510.1 | XP_016867999.1 | 4 | 235 | |||
XM_017012511.1 | XP_016868000.1 | 5 | 487 | |||
XM_017012512.1 | XP_016868001.1 | 4 | 295 | |||
XM_017012513.1 | XP_016868002.1 | 5 | 313 | |||
XM_017012514.1 | XP_016868003.1 | 7 | 725 | |||
XM_017012515.1 | XP_016868004.1 | 6 | 631 | |||
XM_017012516.1 | XP_016868005.1 | 5 | 290 | |||
XM_017012517.1 | XP_016868006.1 | 7 | 709 | |||
XM_017012518.1 | XP_016868007.1 | 8 | 807 | |||
GenBank mRNA | AF402318.1 | - | 6 | 467 | ||
AK021727.1 | - | 6 | 627 | |||
AK291037.1 | - | 6 | 627 | |||
AK299753.1 | - | 5 | 554 | |||
AL833934.1 | - | 3 | 202 | |||
AL834394.1 | - | 6 | 435 | |||
BC017663.1 | - | 6 | 595 | 3883022 | ||
BC063649.1 | - | 8 | 803 |