Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022458.3 | NP_071903.2 | 17 | 2108 | ||
XM_005249555.2 | XP_005249612.1 | 16 | 1921 | |||
XM_005249558.2 | XP_005249615.1 | 12 | 1601 | |||
XM_017012507.1 | XP_016867996.1 | 18 | 2206 | |||
XM_017012508.1 | XP_016867997.1 | 18 | 2164 | |||
XM_017012509.1 | XP_016867998.1 | 17 | 2044 | |||
XM_017012510.1 | XP_016867999.1 | 16 | 1810 | |||
XM_017012511.1 | XP_016868000.1 | 16 | 1939 | |||
XM_017012512.1 | XP_016868001.1 | 16 | 1870 | |||
XM_017012513.1 | XP_016868002.1 | 17 | 1888 | |||
XM_017012514.1 | XP_016868003.1 | 18 | 2177 | |||
XM_017012515.1 | XP_016868004.1 | 13 | 1724 | |||
XM_017012516.1 | XP_016868005.1 | 15 | 1580 | |||
XM_017012517.1 | XP_016868006.1 | 18 | 2161 | |||
XM_017012518.1 | XP_016868007.1 | 19 | 2259 | |||
GenBank mRNA | AF063592.1 | - | 192 | |||
AF348513.1 | - | 4 | 828 | |||
AF402318.1 | - | 17 | 1918 | |||
AK021727.1 | - | 8 | 1294 | |||
AK026940.1 | - | 4 | 834 | |||
AK299753.1 | - | 15 | 1844 | |||
AL833934.1 | - | 13 | 1492 | |||
AL834394.1 | - | 18 | 2010 | |||
BC017663.1 | - | 17 | 2047 | 3883022 | ||
BC063649.1 | - | 19 | 2255 |