Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022473.2 | NP_071918.1 | 2 | 1195 | ||
XM_005254591.2 | XP_005254648.1 | 5 | 1102 | |||
XM_005254592.2 | XP_005254649.1 | 5 | 1102 | |||
XM_017022483.1 | XP_016877972.1 | 4 | 877 | |||
XM_017022484.1 | XP_016877973.1 | 2 | 622 | |||
GenBank mRNA | AF205632.1 | - | 2 | 1168 | ||
AK057410.1 | - | 1 | 176 | |||
AK091929.1 | - | 3 | 931 | |||
AK294529.1 | - | 2 | 925 | |||
BC029366.1 | - | 2 | 1159 | |||
BC058023.1 | - | 2 | 1164 | |||
BC070244.1 | - | 2 | 1159 | |||
BC111396.1 | - | 2 | 1164 | |||
BX648629.1 | - | 1 | 299 | |||
BX649141.1 | - | 5 | 1079 |